1H,1H,2H,2H-Nonafluorohexyl methacrylate 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC5906E-1G PC5906E-25G PC5906E-5G
APOSPC5906E-1GEA 29.2 EUR
APOSPC5906E-1G APOSPC5906E-25G APOSPC5906E-5G
1H,1H,2H,2H-Nonafluorohexyl methacrylate 97%
1H,1H,2H,2H-Nonafluorohexyl methacrylate
Formule: C₁₀H₉F₉O₂
Kookpunt: 60…62 °C/5mmHg
Dichtheid: 1,402 g/cm³ (25 °C)
Vlampunt: 77 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00236094
CAS nummer: 1799-84-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR