5-Fluoro-m-toluonitrile 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC5914-1G PC5914-25G PC5914-5G PC5914-100G
APOSPC5914-1GEA 14.8 EUR
APOSPC5914-1G APOSPC5914-25G APOSPC5914-5G APOSPC5914-100G
5-Fluoro-m-toluonitrile 95%
5-Fluoro-m-toluonitrile
Formule: C₈H₆FN
Smeltpunt: 50…52 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD03094312
CAS nummer: 216976-30-6
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR