3-Fluoro-5-methylphenylacetonitrile 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

PC5919-1G PC5919-5G PC5919-10G
APOSPC5919-1GEA 58.4 EUR
APOSPC5919-1G APOSPC5919-5G APOSPC5919-10G
3-Fluoro-5-methylphenylacetonitrile 97%
3-Fluoro-5-methylphenylacetonitrile
Formule: C₉H₈FN
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD03094317
CAS nummer: 518070-21-8
UN: 3276
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR