1H,1H,12H,12H-Eicosafluoro-1,12-dodecandiol 96%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC5993-25G PC5993-5G PC5993-1G
APOSPC5993-25GEA 322 EUR
APOSPC5993-25G APOSPC5993-5G APOSPC5993-1G
1H,1H,12H,12H-Eicosafluoro-1,12-dodecandiol 96%
1H,1H,12H,12H-Eicosafluoro-1,12-dodecandiol
Formule: C₁₂H₆F₂₀O₂
Kookpunt: 170 °C/5mmHg
Smeltpunt: 156…160 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00236625
CAS nummer: 183162-43-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR