4-Chloro-2-iodo-5-(trifluoromethyl)aniline

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC6061-1G PC6061-500MG PC6061-5G
APOSPC6061-1GEA 220 EUR
APOSPC6061-1G APOSPC6061-500MG APOSPC6061-5G
4-Chloro-2-iodo-5-(trifluoromethyl)aniline
4-Chloro-2-iodo-5-(trifluoromethyl)aniline
Formule: C₇H₄ClF₃IN
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD12827773
CAS nummer: 852569-36-9

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR