1H,1H,2H,2H-Perfluorooctyl methacrylate 99%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC6172E-10G PC6172E-250G PC6172E-50G
APOSPC6172E-10GEA 22 EUR
APOSPC6172E-10G APOSPC6172E-250G APOSPC6172E-50G
1H,1H,2H,2H-Perfluorooctyl methacrylate 99%
1H,1H,2H,2H-Perfluorooctyl methacrylate
Formule: C₁₂H₉F₁₃O₂
Kookpunt: 92 °C/8mmHg
Smeltpunt: –33 °C
Dichtheid: 1,496 g/cm³ (25 °C)
Vlampunt: 113 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00077580
CAS nummer: 2144-53-8
EINECS: 218-407-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR