4-Bromo-2-(trifluoromethyl)benzoic acid

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC6177-1G PC6177-5G PC6177-25G
APOSPC6177-1GEA 22 EUR
APOSPC6177-1G APOSPC6177-5G APOSPC6177-25G
4-Bromo-2-(trifluoromethyl)benzoic acid
4-Bromo-2-(trifluoromethyl)benzoic acid
Formule: C₈H₄BrF₃O₂
Smeltpunt: 116…121 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD07787468
CAS nummer: 320-31-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR