2'-Chloro-4'-(trifluoromethoxy)acetanilide

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC6218-1G PC6218-5G PC6218-25G
APOSPC6218-1GEA 29.2 EUR
APOSPC6218-1G APOSPC6218-5G APOSPC6218-25G
2'-Chloro-4'-(trifluoromethoxy)acetanilide
2'-Chloro-4'-(trifluoromethoxy)acetanilide
Formule: C₉H₇ClF₃NO₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD01571100
CAS nummer: 256529-43-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR