Tris(perfluorobutyl)amine

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: Perfluorotri-n-butylamine, Heptacosafluorotri-n-butylamine, Perfluorotributylamine, FC-43, Heptacosafluorotributylamine, PFTBA, Tris(nonafluorobutyl)amine

PC6220-100G PC6220-25G
APOSPC6220-100GEA 88.2 EUR
APOSPC6220-100G APOSPC6220-25G
Tris(perfluorobutyl)amine
Tris(perfluorobutyl)amine
Formule: C₁₂F₂₇N
MW: 671.1 g/mol
Kookpunt: 170…180 °C
Smeltpunt: -55
Dichtheid: 1.9 g/cm³ (25 °C)
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00000436
CAS nummer: 311-89-7
EINECS: 206-223-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR