(E)-2-(Trifluoromethyl)vinyl benzyl ether

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

PC6259-25G PC6259-5G PC6259-1G
APOSPC6259-25GEA 805 EUR
APOSPC6259-25G APOSPC6259-5G APOSPC6259-1G
(E)-2-(Trifluoromethyl)vinyl benzyl ether
(E)-2-(Trifluoromethyl)vinyl benzyl ether
Formule: C₁₀H₉F₃O
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD13193051
CAS nummer: 932395-40-9
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR