Potassium hexafluoroantimonate 99%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC6279-25G PC6279-100G PC6279-500G PC6279-5G
APOSPC6279-25GEA 111 EUR
APOSPC6279-25G APOSPC6279-100G APOSPC6279-500G APOSPC6279-5G
Potassium hexafluoroantimonate 99%
Potassium hexafluoroantimonate
Formule: F₆KSb
Kookpunt: 1505 °C
Smeltpunt: 846 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00043080
CAS nummer: 16893-92-8
UN: 1549
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR