Potassium tetrafluoroborate 99%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

PC6310-500G
APOSPC6310-500GEA 132 EUR
APOSPC6310-500G
Potassium tetrafluoroborate 99%
Potassium tetrafluoroborate
Formule: BF₄K
Smeltpunt: 530 °C
Dichtheid: 2,505 g/cm³ (25 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011395
CAS nummer: 14075-53-7
EINECS: 237-928-2
UN: 3260
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR