2-Fluoro-3-(trifluoromethoxy)benzyl bromide 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

PC6313-1G PC6313-5G PC6313-25G
APOSPC6313-1GEA 55.5 EUR
APOSPC6313-1G APOSPC6313-5G APOSPC6313-25G
2-Fluoro-3-(trifluoromethoxy)benzyl bromide 95%
2-Fluoro-3-(trifluoromethoxy)benzyl bromide
Formule: C₈H₅BrF₄O
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD12026474
CAS nummer: 1159512-59-0
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR