2-Fluoro-3-(trifluoromethoxy)benzylamine

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

PC6357-1G PC6357-250MG PC6357-100MG
APOSPC6357-1GEA 368 EUR
APOSPC6357-1G APOSPC6357-250MG APOSPC6357-100MG
2-Fluoro-3-(trifluoromethoxy)benzylamine
2-Fluoro-3-(trifluoromethoxy)benzylamine
Formule: C₈H₇F₄NO
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD12026479
CAS nummer: 1159512-65-8
UN: 2735
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR