Sodium chlorodifluoroacetate 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC6460-100G PC6460-25G PC6460-500G
APOSPC6460-100GEA 58.4 EUR
APOSPC6460-100G APOSPC6460-25G APOSPC6460-500G
Sodium chlorodifluoroacetate 97%
Sodium chlorodifluoroacetate
Formule: C₂ClF₂NaO₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00064771
CAS nummer: 1895-39-2
EINECS: 217-586-0

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR