6-Fluoro-4-hydroxycoumarin

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC6507-1G PC6507-250MG
APOSPC6507-1GEA 43.8 EUR
APOSPC6507-1G APOSPC6507-250MG
6-Fluoro-4-hydroxycoumarin
6-Fluoro-4-hydroxycoumarin
Formule: C₉H₅FO₃
Smeltpunt: 248…251 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD03094006
CAS nummer: 1994-13-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR