Sodium hexafluorotitanate 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC6514-5G PC6514-10G PC6514-50G
APOSPC6514-5GEA 73 EUR
APOSPC6514-5G APOSPC6514-10G APOSPC6514-50G
Sodium hexafluorotitanate 98%
Sodium hexafluorotitanate
Formule: F₆Na₂Ti
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00014244
CAS nummer: 17116-13-1
EINECS: 241-181-8

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR