Tetrabutylammonium fluoride trihydrate 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

PC6633T-100G PC6633T-500G PC6633T-1KG
APOSPC6633T-100GEA 22 EUR
APOSPC6633T-100G APOSPC6633T-500G APOSPC6633T-1KG
Tetrabutylammonium fluoride trihydrate 97%
Tetrabutylammonium fluoride trihydrate

Technical bulletin available

Formule: C₁₆H₃₆FN·3H₂O
Smeltpunt: 54…63 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00149981
CAS nummer: 87749-50-6
EINECS: 207-057-2
UN: 1759
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR