3-Fluoropicolinic acid

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC7057-25G PC7057-5G PC7057-1G
APOSPC7057-25GEA 81.8 EUR
APOSPC7057-25G APOSPC7057-5G APOSPC7057-1G
3-Fluoropicolinic acid
3-Fluoropicolinic acid
Formule: C₆H₄FNO₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD04114108
CAS nummer: 152126-31-3

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR