4-Bromo-2-fluorostyrene

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC7069-1G PC7069-5G
APOSPC7069-1GEA 246 EUR
APOSPC7069-1G APOSPC7069-5G
4-Bromo-2-fluorostyrene
4-Bromo-2-fluorostyrene
Formule: C₈H₆BrF
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD09907860
CAS nummer: 627463-17-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR