4'-Chloro-2'-fluoro-5'-methylacetophenone

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: 1-(4-Chloro-2-fluoro-5-methylphenyl)ethanone

PC7084-1G PC7084-250MG
APOSPC7084-1GEA 57.2 EUR
APOSPC7084-1G APOSPC7084-250MG
4'-Chloro-2'-fluoro-5'-methylacetophenone
4'-Chloro-2'-fluoro-5'-methylacetophenone
Formule: C₉H₈ClFO
MW: 186.6106 g/mol
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00236601
CAS nummer: 177211-26-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR