2-Fluoroisonicotinonitrile 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC7107-5G PC7107-25G PC7107-100G
APOSPC7107-5GEA 36.6 EUR
APOSPC7107-5G APOSPC7107-25G APOSPC7107-100G
2-Fluoroisonicotinonitrile 98%
2-Fluoroisonicotinonitrile
Formule: C₆H₃FN₂
Smeltpunt: 29…34 °C
Vlampunt: 82 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD09027299
CAS nummer: 3939-14-8
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR