4-(5-Trifluoromethyl-2-pyridyloxy)benzenesulphonyl chloride

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

PC7227-1G PC7227-5G
APOSPC7227-1GEA 51.2 EUR
APOSPC7227-1G APOSPC7227-5G
4-(5-Trifluoromethyl-2-pyridyloxy)benzenesulphonyl chloride
4-(5-Trifluoromethyl-2-pyridyloxy)benzenesulphonyl chloride
Formule: C₁₂H₇ClF₃NO₃S
MW: 337.697 g/mol
Smeltpunt: 51…53 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD07774162
CAS nummer: 874838-96-7
UN: 3261
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR