2,3,6-Trifluorobenzyl bromide 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: α-Bromo-2,3,6-trifluorotoluene, 2-(Bromomethyl)-1,3,4-trifluorobenzene

PC7285P-25G PC7285P-5G
APOSPC7285P-25GEA 191 EUR
APOSPC7285P-25G APOSPC7285P-5G
2,3,6-Trifluorobenzyl bromide 97%
2,3,6-Trifluorobenzyl bromide
Formule: C₇H₄BrF₃
MW: 225.01 g/mol
Kookpunt: 114 °C
Smeltpunt: 21
Dichtheid: 1.718 g/cm³ (25 °C)
Vlampunt: 91 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00061208
CAS nummer: 151412-02-1
UN: 3265
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR