4-Fluoro-3-(trifluoromethoxy)benzonitrile 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC7297-1G PC7297-5G
APOSPC7297-1GEA 40.9 EUR
APOSPC7297-1G APOSPC7297-5G
4-Fluoro-3-(trifluoromethoxy)benzonitrile 98%
4-Fluoro-3-(trifluoromethoxy)benzonitrile
Formule: C₈H₃F₄NO
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD06660253
CAS nummer: 886501-14-0
UN: 3276
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR