4-Diethylamino-1,1,1-trifluorobut-3-en-2-one

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC7356-10G PC7356-1G PC7356-5G
APOSPC7356-10GEA 461 EUR
APOSPC7356-10G APOSPC7356-1G APOSPC7356-5G
4-Diethylamino-1,1,1-trifluorobut-3-en-2-one
4-Diethylamino-1,1,1-trifluorobut-3-en-2-one
Formule: C₈H₁₂F₃NO
MW: 195.19 g/mol
Kookpunt: 97…98 °C/ 2mmHg
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00115108
CAS nummer: 21045-62-5

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR