2-Bromo-4-fluorobenzyl alcohol 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: 2-Bromo-4-fluorobenzyl Alcohol, (2-Bromo-4-fluorophenyl)methanol

PC7372-25G PC7372-5G
APOSPC7372-25GEA 70.1 EUR
APOSPC7372-25G APOSPC7372-5G
2-Bromo-4-fluorobenzyl alcohol 98%
2-Bromo-4-fluorobenzyl alcohol
Formule: C₇H₆BrFO
MW: 205.03
Smeltpunt: 68…71 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00672925
CAS nummer: 229027-89-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR