1,2-Bis[3-(trifluoromethyl)phenyl]-1,1,2,2-tetrafluoroethane 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC7397-1G PC7397-25G PC7397-5G
APOSPC7397-1GEA 103 EUR
APOSPC7397-1G APOSPC7397-25G APOSPC7397-5G
1,2-Bis[3-(trifluoromethyl)phenyl]-1,1,2,2-tetrafluoroethane 97%
1,2-Bis[3-(trifluoromethyl)phenyl]-1,1,2,2-tetrafluoroethane
Formule: C₁₆H₈F₁₀
Smeltpunt: 82…83 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD01631640
CAS nummer: 287172-67-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR