4-(Trifluoromethoxy)benzoic acid hydrazide 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC7438D-1G PC7438D-25G PC7438D-5G
APOSPC7438D-1GEA 29.2 EUR
APOSPC7438D-1G APOSPC7438D-25G APOSPC7438D-5G
4-(Trifluoromethoxy)benzoic acid hydrazide 95%
4-(Trifluoromethoxy)benzoic acid hydrazide
Formule: C₈H₇F₃N₂O₂
Smeltpunt: 110…120 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00204234
CAS nummer: 175277-18-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR