4-(Trifluoromethoxy)benzyl alcohol 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC7438SD-10G PC7438SD-25G PC7438SD-100G
APOSPC7438SD-10GEA 22 EUR
APOSPC7438SD-10G APOSPC7438SD-25G APOSPC7438SD-100G
4-(Trifluoromethoxy)benzyl alcohol 97%
4-(Trifluoromethoxy)benzyl alcohol
Formule: C₈H₇F₃O₂
Kookpunt: 100…102 °C/12mmHg
Dichtheid: 1,326 g/cm³ (25 °C)
Vlampunt: 98 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00036029
CAS nummer: 1736-74-9

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR