4-(Trifluoromethoxy)phenylhydrazine hydrochloride 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC7439PT-100G PC7439PT-25G PC7439PT-5G
APOSPC7439PT-100GEA 114 EUR
APOSPC7439PT-100G APOSPC7439PT-25G APOSPC7439PT-5G
4-(Trifluoromethoxy)phenylhydrazine hydrochloride 97%
4-(Trifluoromethoxy)phenylhydrazine hydrochloride
Formule: C₇H₈ClF₃N₂O
Smeltpunt: 230 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00053033
CAS nummer: 133115-72-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR