2-(Trifluoromethoxy)thiophenol 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC7439UX-1G PC7439UX-25G PC7439UX-5G
APOSPC7439UX-1GEA 93.5 EUR
APOSPC7439UX-1G APOSPC7439UX-25G APOSPC7439UX-5G
2-(Trifluoromethoxy)thiophenol 95%
2-(Trifluoromethoxy)thiophenol
Formule: C₇H₅F₃OS
Kookpunt: 53…55 °C/15mmHg
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00052320
CAS nummer: 175278-01-0
UN: 2810
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR