1-[2-Nitro-4-(trifluoromethyl)phenyl]-4-methylpiperidine

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC7454-10G PC7454-1G PC7454-5G
APOSPC7454-10GEA 305 EUR
APOSPC7454-10G APOSPC7454-1G APOSPC7454-5G
1-[2-Nitro-4-(trifluoromethyl)phenyl]-4-methylpiperidine
1-[2-Nitro-4-(trifluoromethyl)phenyl]-4-methylpiperidine
Formule: C₁₃H₁₅F₃N₂O₂
Smeltpunt: 38 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD09027734
CAS nummer: 392237-31-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR