2-(Trifluoromethyl)benzenesulphonyl chloride 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: 2-(Trifluoromethyl)benzene-1-sulphonyl chloride

PC7508-100G PC7508-25G PC7508-5G
APOSPC7508-100GEA 198 EUR
APOSPC7508-100G APOSPC7508-25G APOSPC7508-5G
2-(Trifluoromethyl)benzenesulphonyl chloride 98%
2-(Trifluoromethyl)benzenesulphonyl chloride
Formule: C₇H₄ClF₃O₂S
MW: 244.62 g/mol
Kookpunt: 133…135 °C/14mmHg
Dichtheid: 1.585 g/cm³ (25 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00051696
CAS nummer: 776-04-5
UN: 3265
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR