α,α,α-Trifluoro-p-toluonitrile 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC7562-100G PC7562-1KG PC7562-250G PC7562-5KG
APOSPC7562-100GEA 0 EUR
APOSPC7562-100G APOSPC7562-1KG APOSPC7562-250G APOSPC7562-5KG
α,α,α-Trifluoro-p-toluonitrile 98%
α,α,α-Trifluoro-p-toluonitrile
Formule: C₈H₄F₃N
Kookpunt: 183…185 °C
Smeltpunt: 37…41 °C
Dichtheid: 1,278 g/cm³ (25 °C)
Vlampunt: 71 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00001826
CAS nummer: 455-18-5
EINECS: 207-239-1
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR