3-(Trifluoromethyl)cinnamic acid 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC7663-100G PC7663-250G PC7663-25G
APOSPC7663-100GEA 73 EUR
APOSPC7663-100G APOSPC7663-250G APOSPC7663-25G
3-(Trifluoromethyl)cinnamic acid 98%
3-(Trifluoromethyl)cinnamic acid
Formule: C₁₀H₇F₃O₂
Smeltpunt: 132…136 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00004393
CAS nummer: 779-89-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR