3-(Trifluoromethyl)cyclohexanol 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC7668G-1G PC7668G-5G
APOSPC7668G-1GEA 73 EUR
APOSPC7668G-1G APOSPC7668G-5G
3-(Trifluoromethyl)cyclohexanol 97%
3-(Trifluoromethyl)cyclohexanol
Formule: C₇H₁₁F₃O
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00102143
CAS nummer: 454-63-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR