(α,α,α-Trifluoro-3-tolyl)acetic acid 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: (3-(Trifluoromethyl)phenyl)acetic acid, 3-(Trifluoromethyl)phenylacetic acid

PC7699-25G PC7699-100G PC7699-5G
APOSPC7699-25GEA 35.1 EUR
APOSPC7699-25G APOSPC7699-100G APOSPC7699-5G
(α,α,α-Trifluoro-3-tolyl)acetic acid 97%
(α,α,α-Trifluoro-3-tolyl)acetic acid
Formule: (C₆H₄(CF₃))CH₂COOH
MW: 204.15 g/mol
Smeltpunt: 76…79 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00004339
CAS nummer: 351-35-9
EINECS: 206-511-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR