6-Bromo-3-ethoxy-2-fluorobenzeneboronic acid 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC7702-1G PC7702-5G
APOSPC7702-1GEA 125 EUR
APOSPC7702-1G APOSPC7702-5G
6-Bromo-3-ethoxy-2-fluorobenzeneboronic acid 98%
6-Bromo-3-ethoxy-2-fluorobenzeneboronic acid
Formule: C₈H₉BBrFO₃
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD09475871
CAS nummer: 871126-14-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR