3-Bromo-4-(trifluoromethyl)pyridine

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC7718-25G PC7718-5G
APOSPC7718-25GEA 402 EUR
APOSPC7718-25G APOSPC7718-5G
3-Bromo-4-(trifluoromethyl)pyridine
3-Bromo-4-(trifluoromethyl)pyridine
Formule: C₆H₃BrF₃N
Kookpunt: 58 °C/11mmHg
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD08061310
CAS nummer: 936841-70-2
UN: 2810
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR