3-Bromo-2,6-diluorotoluene 99%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: 3-Bromo-2,6-difluorotoluene, 1-Bromo-2,4-difluoro-3-methylbenzene

PC7858-25G PC7858-5G
APOSPC7858-25GEA 95.5 EUR
APOSPC7858-25G APOSPC7858-5G
3-Bromo-2,6-diluorotoluene 99%
3-Bromo-2,6-diluorotoluene
Formule: C₇H₅BrF₂
MW: 207.02 g/mol
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD03412221
CAS nummer: 221220-97-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR