2,4-Difluoro-3-methylbenzonitrile 99%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

PC7906-1G PC7906-5G
APOSPC7906-1GEA 35.1 EUR
APOSPC7906-1G APOSPC7906-5G
2,4-Difluoro-3-methylbenzonitrile 99%
2,4-Difluoro-3-methylbenzonitrile
Formule: C₈H₅F₂N
Smeltpunt: 42…45 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD07777155
CAS nummer: 847502-87-8
UN: 3439
ADR: 6.1,III

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR