Zirconium tetrafluoride 99,5%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Zirconium (IV) fluoride

PC8040-25G PC8040-5G
APOSPC8040-25GEA 91.1 EUR
APOSPC8040-25G APOSPC8040-5G
Zirconium tetrafluoride 99,5%
Zirconium tetrafluoride
Formule: F₄Zr
MW: 167.22 g/mol
Kookpunt: 910 °C
Smeltpunt: 640 °C
Dichtheid: 4.43 g/cm³ (25 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011307
CAS nummer: 7783-64-4
EINECS: 232-018-1
UN: 3260
ADR: 8 II
Merck Index: 12,10308

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR