3-Bromo-2-fluoroaniline 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC8089-5G PC8089-1G PC8089-25G
APOSPC8089-5GEA 26.3 EUR
APOSPC8089-5G APOSPC8089-1G APOSPC8089-25G
3-Bromo-2-fluoroaniline 98%
3-Bromo-2-fluoroaniline
Formule: C₆H₅BrFN
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD09864700
CAS nummer: 58534-95-5
UN: 2810
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR