4-Bromo-2,3,6-trifluorobenzyl alcohol 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC8248-1G PC8248-250MG
APOSPC8248-1GEA 636 EUR
APOSPC8248-1G APOSPC8248-250MG
4-Bromo-2,3,6-trifluorobenzyl alcohol 95%
4-Bromo-2,3,6-trifluorobenzyl alcohol
Formule: C₇H₄BrF₃O
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD03094469
CAS nummer: 252004-34-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR