5-Bromo-1-(2-chloro-6-fluorobenzyl)-6-oxo-1,6-dihydro-3-pyridinecarbaldehyde

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC8570-1G PC8570-500MG
APOSPC8570-1GEA 349 EUR
APOSPC8570-1G APOSPC8570-500MG
5-Bromo-1-(2-chloro-6-fluorobenzyl)-6-oxo-1,6-dihydro-3-pyridinecarbaldehyde
5-Bromo-1-(2-chloro-6-fluorobenzyl)-6-oxo-1,6-dihydro-3-pyridinecarbaldehyde
Formule: C₁₃H₈BrClFNO₂
MW: 344.57 g/mol
Smeltpunt: 148…150 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD09817442
CAS nummer: 952183-68-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR