3-Nitro-5-(trifluoromethyl)aniline

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

PC8673-1G PC8673-25G PC8673-5G
APOSPC8673-1GEA 22 EUR
APOSPC8673-1G APOSPC8673-25G APOSPC8673-5G
3-Nitro-5-(trifluoromethyl)aniline
3-Nitro-5-(trifluoromethyl)aniline
Formule: C₇H₅F₃N₂O₂
Smeltpunt: 77…81 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00061229
CAS nummer: 401-94-5
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR