2-Chloro-N-ethyl-6-fluorobenzylamine

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

PC8700-1G PC8700-250MG
APOSPC8700-1GEA 123 EUR
APOSPC8700-1G APOSPC8700-250MG
2-Chloro-N-ethyl-6-fluorobenzylamine
2-Chloro-N-ethyl-6-fluorobenzylamine
Formule: C₉H₁₁ClFN
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00053426
CAS nummer: 62924-59-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR