Fmoc-4-fluoro-L-phenylalanine 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC8734-1G PC8734-25G PC8734-5G
APOSPC8734-1GEA 62.9 EUR
APOSPC8734-1G APOSPC8734-25G APOSPC8734-5G
Fmoc-4-fluoro-L-phenylalanine 97%
Fmoc-4-fluoro-L-phenylalanine
Formule: C₂₄H₂₀FNO₄
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00191197
CAS nummer: 169243-86-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR