N-[4-(Trifluoromethyl)pyrimidin-2-yl]ethane-1,2-diamine

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC8813-1G PC8813-100MG
APOSPC8813-1GEA 150 EUR
APOSPC8813-1G APOSPC8813-100MG
N-[4-(Trifluoromethyl)pyrimidin-2-yl]ethane-1,2-diamine
N-[4-(Trifluoromethyl)pyrimidin-2-yl]ethane-1,2-diamine
Formule: C₇H₉F₃N₄
Kookpunt: 104…106 °C (0,75 mm Hg)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00816843
CAS nummer: 215655-29-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR